Learn More
Tocris Bioscience™ CHR 6494 trifluoroacetate
Potent and selective haspin inhibitor
Brand: Tocris Bioscience™ 5326/10
1835.81 SEK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
CHR 6494 trifluoroacetate is a potent and selective haspin kinase inhibitor (IC50 = 2 nM). Selective over 27 other protein kinases including Aurora B kinase. Induces cell cycle arrest at G2/M, inhibits cell proliferation and promotes apoptosis in multiple cancer cell lines. Inhibits angiogenesis ex vivo and attenuates tumor growth of HCT-116 xenografts in rats.Specifications
>98% | |
CHR 6494 trifluoroacetate | |
CHR 6494 trifluoroacetate |
10 mg | |
406.36 |
For Research Use Only