Learn More
Macherey-Nagel™ Chromabond™ BT 15 SiOH Column
Brand: Macherey-Nagel™ 732961
This item is not returnable.
View return policy
Specifications
116 mm | |
Flash Column | |
Biotage Isolera Systems |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ BT 15 SiOH Column > I.D.: 12.4mm
Spot an opportunity for improvement?Share a Content Correction