missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab CHROMagar™ Salmonella Plus
CHROMagar Salmonella Plus is chromogenic dehydrated medium for detection and isolation of Salmonella species including lactose positive Salmonella in food specimens.
Brand: Scharlab 00000SA162
This item is not returnable.
View return policy
Description
- Chromogenic medium for the isolation of Salmonella species including S.Typhi and lactose positive Salmonella.
- This medium meets the ISO 6579 : 2002 ISO norm.
- Mainly due to contamination in the food chain and/or during food-production processes, Salmonella commonly induces enteric illness whose major symptoms are abdominal cramps, diarrhea, nausea, and vomiting.
- Second most important foodborne pathogen, its detection remains a big concern in the medical field, and a big challenge in the food industry including the lactose plus strains largely encountered in the poultry.
Specifications
ISO 6579 | |
CHROMagar Salmonella Plus | |
Gram-negative Bacteria | |
Detection and Isolation of Salmonella Species | |
18 to 24 hr. | |
Salmonella |
Dehydrated | |
Dehydrated Culture Media | |
Agar 15.0 g/L, Peptone and Yeast Extract 8.0 g/L, Salts 8.5 g/L, Chromogenic Mix 1.3 g/L, Growth Mix 6.0 g/L | |
37°C | |
7.5 ±0.2 | |
5 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction