Learn More
BD Chromagar™ Salmonella
Brand:  BD 254104
Product Code. 11798972
  • 830.00 SEK / Pack of 20
Estimated Shipment: 15-11-2024
to see stock.
Explore more special offers
Specifications
Salmonella species from clinical stool specimens
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • CHROMagar Salmonella
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P336-Thaw frosted parts with lukewarm water. Do no rub affected area.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : bile extract