missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek RFP-Booster ATTO594

RFP VHH conjugated to ATTO 594.

1150.00 SEK - 3965.00 SEK

Specifications

Clonality Monoclonal
For Use With (Application) Immunofluorescence (IF), Immunohistochemistry (IHC), Immunocytochemistry (ICC)
Conjugate ATTO594
Target Specificity mCherry, mPlum, mRFP, mRFPruby, mKate2
Content And Storage Shipped at ambient temperature. Upon receipt store at +4°C. Stable for 6 month. Do not freeze. Protect from light.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17363433
View Documents Promotion Details
Proteintech
rba594-10
10 μL
1150.00 SEK
10µL
In Stock
to see stock.
Add to basket
 
17373433
View Documents Promotion Details
Proteintech
rba594-100
100 μL
3965.00 SEK
100µL
In Stock
to see stock.
Add to basket
 
Description

Description

  • anti-RFP VHH/ Nanobody conjugated to fluorophore for IF, of mCherry and other red fluorescent protein-fusion proteins
Specifications

Specifications

Monoclonal
ATTO594
Shipped at ambient temperature. Upon receipt store at +4°C. Stable for 6 month. Do not freeze. Protect from light.
Immunofluorescence (IF), Immunohistochemistry (IHC), Immunocytochemistry (ICC)
mCherry, mPlum, mRFP, mRFPruby, mKate2
Videos
SDS
Documents

Documents

Product Certifications