missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ciclopirox olamine

Product Code. 16504244
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41621-49-2
Molecular Formula C14H24N2O3
Molecular Weight (g/mol) 268.36
InChI Key MBRHNTMUYWQHMR-UHFFFAOYSA-N
IUPAC Name 2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one
SMILES NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1

Specifications

Melting Point 143.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: .. Other solubilities: almost insoluble in dimethylsulfoxide and methanol
Formula Weight 268.35
Chemical Name or Material Ciclopirox olamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.