missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Amino-1-cyclohexanecarboxamide, 98+%, Thermo Scientific Chemicals

481.62 SEK - 1810.61 SEK

Chemical Identifiers

CAS 115014-77-2
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.2
MDL Number MFCD00143979
InChI Key DXNKJRKPVQVDJW-WDSKDSINSA-N
Synonym cyclohexanecarboxamide,2-amino-, 1r,2s-rel, 1s-2beta-aminocyclohexane-1alpha-carboxamide
PubChem CID 30069041
IUPAC Name (1S,2S)-2-aminocyclohexane-1-carboxamide
SMILES C1CCC(C(C1)C(=O)N)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10501521
SDS View Documents
Thermo Scientific Acros
277772500
250 mg N/A N/A N/A
10482825
SDS View Documents
Thermo Scientific Acros
277770010
1 g N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
cis-2-Amino-1-cyclohexanecarboxamide, 98+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.