Learn More
cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals
5940.00 SEK - 19995.00 SEK
Chemical Identifiers
CAS | 685508-28-5 |
---|---|
Molecular Formula | C13H15NO4 |
Molecular Weight (g/mol) | 249.266 |
MDL Number | MFCD03844615 |
InChI Key | ZNRUEEQIUVKKBL-MNOVXSKESA-N |
Synonym | cis-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, cis-2-bocamino cyclobutanecarboxylic acid, cis-2-cbz-amino cyclobutane-1-carboxylic acid, 2beta-benzyloxycarbonylamino cyclobutane-1beta-carboxylic acid, 1r,2s-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid |
PubChem CID | 68498468 |
IUPAC Name | (1R,2S)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid |
SMILES | C1CC(C1C(=O)O)NC(=O)OCC2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15417296
|
Thermo Scientific Alfa Aesar
H62329.MD |
250 mg |
5940.00 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407296
|
Thermo Scientific Alfa Aesar
H62329.03 |
1 g |
19995.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NotesStore in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
685508-28-5 | |
249.266 | |
ZNRUEEQIUVKKBL-MNOVXSKESA-N | |
68498468 | |
C1CC(C1C(=O)O)NC(=O)OCC2=CC=CC=C2 |
C13H15NO4 | |
MFCD03844615 | |
cis-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, cis-2-bocamino cyclobutanecarboxylic acid, cis-2-cbz-amino cyclobutane-1-carboxylic acid, 2beta-benzyloxycarbonylamino cyclobutane-1beta-carboxylic acid, 1r,2s-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid | |
(1R,2S)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid |
Specifications
685508-28-5 | |
MFCD03844615 | |
ZNRUEEQIUVKKBL-MNOVXSKESA-N | |
(1R,2S)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid | |
68498468 | |
97% |
C13H15NO4 | |
cis-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, cis-2-bocamino cyclobutanecarboxylic acid, cis-2-cbz-amino cyclobutane-1-carboxylic acid, 2beta-benzyloxycarbonylamino cyclobutane-1beta-carboxylic acid, 1r,2s-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid | |
C1CC(C1C(=O)O)NC(=O)OCC2=CC=CC=C2 | |
249.266 | |
249.26 | |
cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.