missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Hexen-1-ol, 94%, remainder mainly trans isomer, Thermo Scientific Chemicals

Product Code. 11474754
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-94-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00063209
InChI Key ZCHHRLHTBGRGOT-PLNGDYQASA-N
Synonym cis-2-hexen-1-ol, 2-hexen-1-ol, z, cis-hex-2-en-1-ol, z-2-hexen-1-ol, z-hex-2-en-1-ol, unii-871tel510e, 2z-hexen-1-ol, 2z-hex-2-en-1-ol, cis-1-hydroxy-2-hexene, cis-2-hexenol
PubChem CID 5324489
IUPAC Name (Z)-hex-2-en-1-ol
SMILES CCC\C=C/CO

Specifications

Density 0.847
Boiling Point 158°C
Flash Point 54°C (129°F)
Refractive Index 1.441
Quantity 5 g
UN Number UN1987
Beilstein 1719708
Formula Weight 100.16
Percent Purity 94%
Assay remainder mainly trans isomer
Chemical Name or Material cis-2-Hexen-1-ol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
cis-2-Hexen-1-ol, 94%, remainder mainly trans isomer, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.