Learn More
RETSCH Clamping Covers
Brand:  RETSCH 02.660.0012
6061.00 SEK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10501075
  • 6380.00 SEK / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Specifications
Clamping lid with large window of Perspex
Test Sieves 400 mm dia.
1 Ea.