Learn More
Kartell™ Clamps for Glass Joints
Plastic clamps for safe connection of standard-finish glass conical joints
Brand:  Kartell™ 3454000
Additional Details : Weight : 0.23400kg
 View more versions of this product
Product Code. 15835587
  • 800.00 SEK / Pack of 10
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Description
Description
  • Corrosion proof
  • Do not scratch glass
  • Available in various colours according to dimension
  • Autoclavable
Specifications
Specifications
10/Pk.
POM
45/40
Brown