Learn More
Abeba™ Classic 1001 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
Brand:  Abeba™ 1001/43
 View more versions of this product
Product Code. 15626577
  • 990.00 SEK / 1 pair
Estimated Shipment: 05-05-2025
to see stock.
Explore more special offers
Description
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Microfiber
  • White
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Microfiber lining and heel patch
  • Adjustable instep area
  • Buckle fastening
Specifications
Specifications
CE EN ISO 20345:2011 SB A, E, FO SRC
Microfiber
43
Polyurethane
Steel
White
Microfiber
Slip-resistant
1001
Animal fats, oil and fuel
Product Title
Abeba™ Classic 1001 Shoes > Size: 43