Learn More
Abeba™ Clean Room 3620 Shoes
Special ESD clean room shoes.
1090.00 SEK
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E SRA |
---|---|
Color | White |
Material | Microfiber |
Style | 3620 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15600378
|
Abeba™
3620/35 |
35 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15610378
|
Abeba™
3620/36 |
36 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15620378
|
Abeba™
3620/37 |
37 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15630378
|
Abeba™
3620/38 |
38 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15640378
|
Abeba™
3620/39 |
39 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15650378
|
Abeba™
3620/40 |
40 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15660378
|
Abeba™
3620/41 |
41 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15670378
|
Abeba™
3620/42 |
42 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15680378
|
Abeba™
3620/43 |
43 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15690378
|
Abeba™
3620/44 |
44 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15600388
|
Abeba™
3620/45 |
45 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15610388
|
Abeba™
3620/46 |
46 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15620388
|
Abeba™
3620/47 |
47 |
1090.00 SEK
1 pair |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E SRA
- Microfiber
- White
- ESD dissipative according to DIN EN 61340
- Fixed gaitor, cleanroom textile
Specifications
CE EN ISO 20347:2012 OB A, E SRA | |
White | |
Microfiber | |
3620 |