missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clemastine fumarate, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
H1-receptor antagonist. Enhances differentiation of oligodendrocyte progenitor cells into myelin basic protein-positive oligodendrocytes in vitro and promotes remyelination in vivo. Also displays antimuscarinic properties. Clinically-used antihistamine.
Specifications
Specifications
CAS | 14976-57-9 |
Molecular Formula | C25H30ClNO5 |
Quantity | 100 mg |
Synonym | clemastine fumarate, agasten, mecloprodine, aloginan, xolamin, tavist, alphamin, anhistan, clemanil, fuluminol |
InChI Key | PMGQWSIVQFOFOQ-YKVZVUFRSA-N |
SMILES | CC(C1=CC=CC=C1)(C2=CC=C(C=C2)Cl)OCCC3CCCN3C.C(=CC(=O)O)C(=O)O |
IUPAC Name | (E)-but-2-enedioic acid;(2R)-2-[2-[(1R)-1-(4-chlorophenyl)-1-phenylethoxy]ethyl]-1-methylpyrrolidine |
Molecular Weight (g/mol) | 459.967 |
PubChem CID | 5281069 |
ChEBI | CHEBI:3739 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'