Learn More
Nickel Electro™ Clifton™ Stainless Steel Student Spirometer
Nickel Electro™ Clifton™ Stainless Steel Student Spirometer can be used for investigating tidal volumes, respiratory rate, pulmonary ventilation, reserve volumes and vital capacity.
Brand:  Nickel Electro™ SM-1
Product Code. 13122191
  • 18475.00 SEK / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Description
  • A simple lever control enables the subject to be connected either to atmosphere or to the float for recording.
  • A container for soda-lime crystals is fitted which can be used to absorb carbon dioxide.
  • The spirometer is designed to offer minimum resistance to air flow.
  • This means greater comfort for the subject and more realistic results.
  • The tank is one piece stainless steel pressing housed in a durable painted outer case.
  • The float is constructed from lightweight plastic.
  • The float is calibrated 0 to 7 x 0.5 litres and has a metal counterpoise.
  • The pen arm corresponds to approximately 0.5 litres.
  • Supplied with a mouth piece that fits into the corrugated breathing hoses which snugly connects into Spirometer.
  • Also includes a nose clip and pen arm for fitting data logger sensors.
Specifications
Specifications
Included a nose clip and pen arm for fitting data logger sensors.
1/Ea.
Stainlea Steel, Plastic