Learn More
STEMCELL Technologies ClonaCell™-CHO ACF Medium
Serum-free, animal component-free semi-solid medium with supplements for selection and cloning of transfected CHO cells
Brand:  STEMCELL Technologies 03816
Additional Details : Weight : 0.09000kg
Product Code. 17108211
  • 11962.50 SEK / 90mL
Estimated Shipment: 09-08-2024
to see stock.
Description
Description
  • ClonaCell™-CHO ACF is a methylcellulose-based semi-solid medium with supplements for the selection and cloning of suspension-adapted CHO cells in serum-free conditions
  • This medium is animal component-free and contains only recombinant proteins and synthetic components
  • Single-cell cloning efficiency of CHO cells is higher in this medium than in protein-free media, especially at low plating densities
  • It does not contain L-glutamine, selection agents, or phenol red and is compatible with dihydrofolate reductase (DHFR) and glutamine synthase (GS) selection systems
Specifications
Specifications
CHO
Cell Culture; Semi-Solid Cloning
90 mL