Learn More
STEMCELL Technologies ClonaCell™-TCS Medium
Semi-solid methylcellulose-based medium for selection and cloning of suspension-adapted and adherent cells (serum-containing)
Brand:  STEMCELL Technologies 03814
Additional Details : Weight : 0.08000kg
Product Code. 17198201
  • 9699.00 SEK / 80mL
Estimated Shipment: 28-08-2024
to see stock.
Description
Description
  • ClonaCell™-TCS Medium is a serum-containing methylcellulose-based semi-solid medium that can be used for selecting and cloning a variety of suspension-adapted cell lines, including CHO-S and hybridomas
  • It can also be used for semi-solid cloning of some cell lines that grow adherently in the presence of serum, including CHO-K1, BHK-1, and HEK-293
  • This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA) and supports robust growth of a wide variety of cell types
  • ClonaCell™-TCS Medium does not contain selection agents
Specifications
Specifications
Hybridomas; create dimCellLine - CHO Cells, HEK-293
Cell Culture; Semi-Solid Cloning
80 mL