missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Clopidogrel hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective, high affinity P2Y12 receptor antagonist (pKi = 6.9 for hP2Y12 receptor). Inhibits ADP-induced platelet aggregation; potent antithrombotic agent. Requires bioactivation, unlike PSB 0739 .
Active Enantiomer also available.
Specifications
Quantity | 10 mg |
Formula Weight | 358.28 |
Percent Purity | >99% |
Chemical Name or Material | (±)-Clopidogrel hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'