missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTNAP5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5122083
This item is not returnable.
View return policy
Description
May play a role in the correct development and proper functioning of the peripheral and central nervous system and be involved in cell adhesion and intercellular communication.Specifications
CNTNAP5 | |
Polyclonal | |
Unconjugated | |
CNTNAP5 | |
CASPR5; cell recognition molecule Caspr5; CNTNAP5; contactin associated protein like 5; contactin associated protein-like 5; contactin-associated protein-like 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
129684 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q8WYK1 | |
CNTNAP5 | |
Ala25-Asp302 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction