missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobaltous Ammonium Sulfate, MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
Specifications
CAS | 13596-46-8 |
Quantity | 100 g |
Synonym | diammonium cobalt ii sulfate hexahydrate, cobalt 2+ diammonium hexahydrate disulfate, cobaltous ammonium sulfate hexahydrate, sulfuric acid, ammoniumcobalt 2+ salt 2:2:1 |
Formula Weight | 395.23 |
Chemical Name or Material | Cobaltous Ammonium Sulfate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'