Learn More
Palintest™ COD Reagent Test Tubes
These COD reagent test tubes, with a range of 0 to 2000mg/L, are ideal for use in most laboratory and scientific environments where measuring COD.
1075.00 SEK - 1805.00 SEK
Product Code | Brand | Range | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Description | Price | Quantity & Availability | ||||
11918437
|
Palintest™
PL454 |
0 to 2000mg/L | Mercury Free |
1075.00 SEK
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11946473
|
Palintest™
PL464 |
0 to 2000mg/L | With Mercury |
1805.00 SEK
Pack of 24 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11928437
|
Palintest™
PL452 |
0 to 400mg/L | — |
1075.00 SEK
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Ready to use reagent test tubes
- COD reagent test tubes
- Quantity: 1
- Number of Tests: 25
- Range: 0 to 2000mg/L
- Type: COD reagent test tubes
Specifications
25 | |
1 Each | |
COD Reagent Test Tube |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- COD 2000 Solution
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P391-Collect spillage.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, silver sulphate, potassium dichromate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Palintest™ COD Reagent Test Tubes