missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL7A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
1730.00 SEK - 4375.00 SEK
Specifications
Antigen | COL7A1 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17783474
|
Invitrogen
PA5122080 |
20 μL |
1730.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17773474
|
Invitrogen
PA5122079 |
100 μL |
4375.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes the alpha chain of type VII collagen. The type VII collagen fibril, composed of three identical alpha collagen chains, is restricted to the basement zone beneath stratified squamous epithelia. It functions as an anchoring fibril between the external epithelia and the underlying stroma. Mutations in this gene are associated with all forms of dystrophic epidermolysis bullosa. In the absence of mutations, however, an acquired form of this disease can result from an autoimmune response made to type VII collagen. [provided by RefSeq, Jul 2008].Specifications
COL7A1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q02388 | |
1294 | |
Pro190-Asp472 (Accession Q02388), with N-terminal His Tag | |
Primary | |
-20°C | |
COL7A1 |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
alpha-1 type VII collagen; AW209154; COL7A1; collagen alpha-1(VII) chain; collagen type VII alpha 1; collagen type VII alpha 1 chain; collagen vii; collagen VII, alpha-1 polypeptide; collagen, type VII, alpha 1; EBD1; EBDCT; EBR1; HGNC:2214; LC collagen; long chain collagen; long-chain collagen; NDNC8; procollagen, type VII, alpha 1 | |
COL7A1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title