Learn More
Shamrock™ Color-coded Labeling Tape, 12m
Safe for use in autoclaves, freezers, and ovens. Shamrock™ Color-coded Labeling Tape can be written on with marker, ballpoint and felt-tip pen, and even pencil.
71.82 SEK - 134.00 SEK
Specifications
Autoclavable | Autoclavable |
---|---|
Adhesive Type | Removable |
Product Type | Label Tape |
For Use With (Application) | Ideal for Universal Marking Pen, a Pencil and Felt |
For Use With (Equipment) | For Autoclave, Freezer, Fridge and Oven |
Product Code | Brand | Color | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Width (Metric) | Price | Quantity & Availability | ||||
11750075
|
Shamrock™
ST-34-12 |
Violet | 19 mm |
105.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11705493
|
Shamrock™
ST-34-5 |
Orange | 19 mm |
105.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11775483
|
Shamrock™
ST-34-2 |
Yellow | 19 mm |
105.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11795483
|
Shamrock™
ST-34-4 |
Red | 19 mm |
105.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11715493
|
Shamrock™
ST-34-6 |
Blue | 19 mm |
105.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11725753
|
Shamrock™
ST-10-4 |
Red | 25 mm |
134.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11745753
|
Shamrock™
ST-10-6 |
Blue | 25 mm |
134.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11724794
|
Shamrock™
ST-12-2 |
Yellow | 13 mm |
79.20 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11794804
|
Shamrock™
ST-12-3 |
Green | 13 mm |
71.82 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11765483
|
Shamrock™
ST-34-1 |
White | 19 mm |
105.00 SEK
12m |
In Stock
Log in to see stock. |
||||
11705513
|
Shamrock™
ST-34-7 |
Pink | 19 mm |
105.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11745483
|
Shamrock™
ST-12-6 |
Blue | 13 mm |
79.20 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11785483
|
Shamrock™
ST-34-3 |
Green | 19 mm |
105.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11720075
|
Shamrock™
ST-12-12 |
Violet | 13 mm |
79.20 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11794784
|
Shamrock™
ST-12-1 |
White | 13 mm |
79.20 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11725483
|
Shamrock™
ST-12-4 |
Red | 13 mm |
79.20 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11735483
|
Shamrock™
ST-12-5 |
Orange | 13 mm |
79.20 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11755483
|
Shamrock™
ST-12-7 |
Pink | 13 mm |
83.35 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11715513
|
Shamrock™
ST-10-1 |
White | 25 mm |
134.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11735513
|
Shamrock™
ST-10-3 |
Green | 25 mm |
134.00 SEK
12m |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11725513
|
Shamrock™
ST-10-2 |
Yellow | 25 mm |
133.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11780075
|
Shamrock™
ST-10-12 |
Violet | 25 mm |
134.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11735753
|
Shamrock™
ST-10-5 |
Orange | 25 mm |
134.00 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11730075
|
Shamrock™
ST-12-14 |
Gray | 13 mm |
79.20 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11760075
|
Shamrock™
ST-34-14 |
Gray | 19 mm |
94.60 SEK
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Resistant to water, oil and acid
- Temperature -20 to +121°C
- For autoclave, freezer, refrigerator and oven
- Marking with a ballpoint pen, pencil and felt-tip pen
Specifications
Autoclavable | |
Label Tape | |
For Autoclave, Freezer, Fridge and Oven | |
-20°C to +121°C |
Removable | |
Ideal for Universal Marking Pen, a Pencil and Felt | |
1 Ea. | |
12 m |