Learn More
Shamrock™ Color Coded Multi-Purpose 3 Inch Labeling Tape
Offered in a number of colors to fit your needs. Shamrock™ Color Coded Multi-Purpose 3 Inch Labeling Tape offers multiple opportunities for color coding in the laboratory - test, physician or department.
204.00 SEK - 565.00 SEK
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
11879080
|
Shamrock™
ST-120-26 |
Black | 55 m x 13 mm |
204.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11731636
|
Shamrock™
ST-200-6 |
Blue | 55 m x 50 mm |
565.00 SEK
55m |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11827203
|
Shamrock™
ST-340-17 |
Chartreuse | 55 m x 19 mm |
305.00 SEK
55m |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11710095
|
Shamrock™
ST-100-23 |
Brown | 55 m x 25 mm |
356.00 SEK
55m |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11770085
|
Shamrock™
ST-120-23 |
Brown | 55 m x 13 mm |
204.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11852592
|
Shamrock™
ST-100-10 |
Copper | 55 m x 25 mm |
356.00 SEK
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11700095
|
Shamrock™
ST-340-23 |
Dark Brown | 55 m x 19 mm |
277.00 SEK
55m |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11771616
|
Shamrock™
ST-110-29 |
Green, Fluorescent | 55 m x 38 mm |
484.00 SEK
55m |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11772255
|
Shamrock™
ST-200-27 |
Red, Fluorescent | 55 m x 50 mm |
565.00 SEK
55m |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11790085
|
Shamrock™
ST-340-14 |
Gray | 55 m x 19 mm |
277.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11831573
|
Shamrock™
ST-200-13 |
Lavender | 55 m x 50 mm |
565.00 SEK
55m |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11701636
|
Shamrock™
ST-200-3 |
Green | 55 m x 50 mm |
565.00 SEK
55m |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11730454
|
Shamrock™
ST-100-5 |
Orange | 55 m x 25 mm |
356.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11720085
|
Shamrock™
ST-120-14 |
Gray | 55 m x 13 mm |
204.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11897193
|
Shamrock™
ST-340-13 |
Lavender | 55 m x 19 mm |
277.00 SEK
55m |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11721636
|
Shamrock™
ST-200-5 |
Orange | 55 m x 50 mm |
565.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11740414
|
Shamrock™
ST-120-7 |
Pink | 55 m x 13 mm |
204.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11750454
|
Shamrock™
ST-100-7 |
Pink | 55 m x 25 mm |
356.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16377205
|
Shamrock™
ST-110-4 |
Red | 55 m x 38 mm |
484.00 SEK
55m |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11795753
|
Shamrock™
ST-120-4 |
Red | 55 m x 24 mm |
225.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11711636
|
Shamrock™
ST-200-4 |
Red | 55 m x 50 mm |
565.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11710085
|
Shamrock™
ST-120-12 |
Violet | 55 m x 13 mm |
204.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11857153
|
Shamrock™
ST-100-12 |
Violet | 55 m x 25 mm |
356.00 SEK
Each |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11862592
|
Shamrock™
ST-200-11 |
Rose | 55 m x 50 mm |
565.00 SEK
Each |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11770434
|
Shamrock™
ST-100-1 |
White | 55 m x 25 mm |
356.00 SEK
55m |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11751616
|
Shamrock™
ST-110-1 |
White | 55 m x 38 mm |
484.00 SEK
55m |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11791626
|
Shamrock™
ST-200-2 |
Yellow | 55 m x 50 mm |
565.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11781626
|
Shamrock™
ST-200-1 |
White | 55 m x 50 mm |
565.00 SEK
55m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Descrizione
- Resistant to water, oil and acid
- Temperature range of -20 to +121°C
- For autoclave, freezer, refrigerator and oven
- Universal marking with ballpoint pen, paper pencil and felt marker
Specifica
Removable | |
55 m |