Learn More
Thermo Scientific™ Columbia CNA Aesculin Selective Agar / MacConkey Agar No. 3 Modified
Isolate and cultivate staphylococci and streptococci while isolating and detecting Salmonella and Shigella, all on a single plate with two separate media formulations.
Brand: Thermo Scientific™ PB5224E
164.34 SEK valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Isolate and cultivate fastidious microorganisms with clearly visible haemolytic reactions (staphylococci and streptococci), and isolate and detect Salmonella and Shigella spp. using Thermo Scientific™ Columbia CNA Aesculin Selective Agar / MacConkey Agar No. 3 Modified, a single bi-plate which includes two separate media formulations.
Read more results from a single patient sample and produce more diagnostic details of pathogenic organisms while reducing storage capacity issues in the incubator with a bi-plate that includes two widely used microbiological media.Identify both Gram-positive and Gram-negative organisms from one patient sample, including Gram-positive cocci, Enterobacteriaceae, and Acinetobacter species.
On one side of the plate, Columbia CNA Agar with Asculin isolates Gram-positive organisms from clinical samples with this selective microbiological medium traditionally used to identify staphylococci and streptococci because it provides clear haemolysis and typical growth for S. pneumoniae (dent morphology).
On the other side of the plate, MacConkey Agar No. 3 is a selective medium for differentiating between coliforms and lactose and non-lactose fermenters while gram-positive cocci are completely inhibited.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
CAN: Flame red, opaque. MAC3: Antique pink, transparent | |
Columbia CNA Aesculin Selective Agar / MacConkey Agar No. 3 Modified | |
Germany | |
17g ± 5% | |
10/Pk. |
2° to 12°C, in the dark | |
90mm biplate | |
Agar | |
Flow Wrapped - Single Layer |