missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for HU13 Midi Horizontal Gel Electrophoresis Unit, 1mm thick
For use with HU13 Midi Horizontal Gel Electrophoresis Unit
Brand: Fisherbrand™ HU13-C1-10
This item is not returnable.
View return policy
Description
Color coded
Height adjustable
Offer complete control over loading volume
Well depth to maximum 112-sample throughput
Specifications
HU13 Midi Horizontal Gel Electrophoresis Unit | |
10 | |
40 μL |
Comb | |
1 mm | |
9.5 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction