missing translation for 'onlineSavingsMsg'
Learn More

Complement C6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

1935.00 SEK - 4555.00 SEK

Specifications

Antigen Complement C6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18343245
View Documents
Bio-Techne
NBP3-03587-20UL
20 μL
1935.00 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
18337384
View Documents
Bio-Techne
NBP3-03587-100UL
100 μL
4555.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Complement C6 Polyclonal specifically detects Complement C6 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Complement C6
Polyclonal
Purified
RUO
Human, Mouse
complement component 6, complement component C6
Recombinant fusion protein containing a sequence corresponding to amino acids 22-260 of human Complement C6 (NP_000056.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
Rabbit
Immunology, Innate Immunity
PBS with 50% glycerol, pH7.3.
729
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.