missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C5aR1 Rabbit anti-Human, Bovine, Ferret, Rabbit, Clone: S5/1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-12083-0.2MG
This item is not returnable.
View return policy
Description
Complement Component C5aR1 Monoclonal antibody specifically detects Complement Component C5aR1 in Human, Bovine, Ferret, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, NeutralizationSpecifications
Complement Component C5aR1 | |
Monoclonal | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Block/Neutralize | |
C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5a receptor 1 | |
This antibody was raised by immunizing mice with a synthetic peptide corresponding to C5a-R; amino acids: 1-31 (N terminal). | |
0.2 mg | |
GPCR, Immunology, Innate Immunity | |
728 | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Flow Cytometry | |
S5/1 | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction