Learn More
Cytiva Complete End Piece for Use With XK26 Columns
Brand: Cytiva 18-6489-01
This item is not returnable.
View return policy
Description
Specifications
XK26 Columns | |
Ethylene propylene diene (EPDM); Polytetrafluoroethene (PTFE); Acetal plastic (POM) | |
Complete End Piece | |
Locking ring, Washer, Sealing ring, End piece, O-ring (44.12 x 2.62 mm) |
Product Content Correction
Product Title
Cytiva Complete End Piece for Use With XK26 Columns > Complete End Piece for Use With XK26 Columns
Spot an opportunity for improvement?Share a Content Correction