Learn More
Conductivity solution 100,000μS/cm, Fisher chemical™
Click to view available options
Quantity:
500 mL
1000 mL
Packaging:
Bottle
Specifications
Specifications
Packaging | Bottle |
Quantity | 500 mL |
Conductivity | 100,000μS/cm at 25°C |
Type | Conductivity Solution |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1044033 | Certificate of Analysis | 04/03/2025 | CTE 016 500 |
Product Content Correction
Product Title
Conductivity solution 100,000μS/cm, Fisher chemical™ >
Spot an opportunity for improvement?Share a Content Correction