missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5000 μS/cm Conductivity Standard Sachets, Hanna Instruments™
Click to view available options
Quantity:
25 x 20 mL
Packaging:
Sachets
Description
- Premium quality 5,000 μS/cm conductivity solution that is NIST traceable and supplied with a Certificate of Analysis (COA)
- Conductivity solution standardized with a conductivity meter
- Calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) in deionized water for analytical use in accordance with ISO 3696/BS3978
- Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers
- Ideal for users that do not calibrate frequently
- Ideal for field use due to portability
Specifications
Specifications
Packaging | Sachets |
Quantity | 25 x 20 mL |
Color | Colorless |
Odor | Odorless |
pH | 7 |
Solubility | Soluble |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction