missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 30 Polyclonal Antibody (Z-PP9), Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 712200
This item is not returnable.
View return policy
Description
This antibody is specific for the Connexin 30. Cross-reactivity with the highly related Connexin 26 protein or with other Connexin family members has not been detected. Reactivity of this antibody has been confirmed by western blotting using cell lysates derived from adult rat, mouse, human, and cat brains.
Connexin 30 Polyclonal antibody specifically detects Connexin 30 in Feline, Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Connexin 30 | |
Polyclonal | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
O95452, P70689 | |
GJB6 | |
A synthetic peptide derived from the unique C-terminus of the mouse Connexin 30 protein | |
50 μg | |
Primary | |
Feline, Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
Z-PP9 | |
Unconjugated | |
GJB6 | |
AA958971; connexin 30; connexin-30; CX30; Cxn-30; D14Bwg0506e; DFNA3; DFNA3B; DFNB1B; ECTD2; ectodermal dysplasia 2, hidrotic (Clouston syndrome); ED2; EDH; Gap junction beta-6 protein; gap junction membrane channel protein beta 6; gap junction protein beta 6; gap junction protein, beta 6; gap junction protein, beta 6 (connexin 30); gap junction protein, beta 6, 30kDa; Gjb6; HED; HED2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
101082796, 10804, 14623, 84403 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction