Learn More
Cole Parmer CoorsTek High Alumina High-Form Crucible
CoorsTek High Alumina High-Form Crucible is manufactured from 99.8% pure aluminum oxide. Highly resistant to chemicals, alkalines, and other fluxes.
Brand:  Cole Parmer 17804-07
Additional Details : Weight : 0.80000kg
Product Code. 11735479
  • 2545.00 SEK / Each
Estimated Shipment: 10-10-2024
to see stock.
Add to basket
Description
Description
CoorsTek High Alumina High-Form Crucible is manufactured from 99.8% pure aluminum oxide. Highly resistant to chemicals, alkalines, and other fluxes.
  • Manufactured from 99.8% pure aluminum oxide
  • Inactive in hydrogen and carbonaceous atmospheres
  • Crucible covers sold separately
  • Withstands temperatures to 3182°F (1750°C)
Specifications
Specifications
High Alumina
100 mL
58 mm
Crucible
1750°C
Yes
58 mm