Learn More
Copanlisib, MedChemExpress
MedChemExpress Copanlisib (BAY 80-6946) is a potent, selective and ATP-competitive pan-class I PI3K inhibitor, with IC50s of 0.5 nM, 0.7 nM, 3.7 nM and 6.4 nM for PI3Kα, PI3Kδ, PI3Kβ and PI3Kγ, respectively. Copanlisib has more than 2,000-fold selectivity against other lipid and protein kinases, except for mTOR. Copanlisib has superior antitumor activity.

964.76 SEK - 7235.68 SEK
Specifications
Chemical Name or Material | Copanlisib |
---|---|
CAS | 1032568-63-0 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C23H28N8O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16472567
![]() |
Medchem Express
HY-15346 |
50 mg |
4341.41 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16449057
![]() |
Medchem Express
HY-15346 |
5 mg |
964.76 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16436468
![]() |
Medchem Express
HY-15346 |
10 mg |
1447.13 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16426746
![]() |
Medchem Express
HY-15346 |
100 mg |
7235.68 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Copanlisib (BAY 80-6946) is a potent, selective and ATP-competitive pan-class I PI3K inhibitor, with IC50s of 0.5 nM, 0.7 nM, 3.7 nM and 6.4 nM for PI3Kα, PI3Kδ, PI3Kβ and PI3Kγ, respectively. Copanlisib has more than 2,000-fold selectivity against other lipid and protein kinases, except for mTOR. Copanlisib has superior antitumor activity.Specifications
Copanlisib | |
Light Yellow | |
C23H28N8O4 | |
1M HCl : 100 mg/mL (208.11 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.08 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.39 mM; Need ultrasonic) | |
480.52 | |
98.02% | |
Solid |
1032568-63-0 | |
Research | |
BAY 80-6946 | |
O=C(C1=CN=C(N)N=C1)NC2=NC3=C(OC)C(OCCCN4CCOCC4)=CC=C3C5=NCCN25 | |
480.52 | |
Research | |
Cancer-Kinase/protease |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.