missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

423.14 SEK - 2714.45 SEK

Chemical Identifiers

CAS 19004-19-4
Molecular Formula 2·5 H2O
Molecular Weight (g/mol) 232.6
MDL Number MFCD02253001
Synonym dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10413655
View Documents
Thermo Scientific Acros
405851000
100 g Plastic bottle
423.14 SEK
100g
Please to purchase this item. Need a web account? Register with us today!
10177410
View Documents
Thermo Scientific Acros
405855000
500 g Plastic bottle
1585.34 SEK
500g
Please to purchase this item. Need a web account? Register with us today!
10626794
View Documents
Thermo Scientific Acros
405850025
2.5 kg Plastic bottle
2714.45 SEK
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19004-19-4
232.6
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate
2·5 H2O
MFCD02253001

Specifications

114.0°C
Blue
98.0 to 102% (Iodometry)
2·5H2O
17,366
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate
232.6
≥98%
Copper(II) nitrate, hemipentahydrate
19004-19-4
Crystalline Powder
2·5 H2O
MFCD02253001
10, 2633
Solubility in water: soluble
232.6
ACS Reagent
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

Product Identifier
  • Copper(II) nitrate, 2.5-hydrate, reagent ACS

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.