Learn More
Copper(II) oxide, 99.9995% (metals basis), Thermo Scientific Chemicals
1088.71 SEK - 11265.74 SEK
Chemical Identifiers
CAS | 1317-38-0 |
---|---|
Molecular Formula | CuO |
Molecular Weight (g/mol) | 79.55 |
MDL Number | MFCD00010979 |
InChI Key | KKCXRELNMOYFLS-UHFFFAOYSA-N |
Synonym | cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide |
PubChem CID | 14829 |
IUPAC Name | oxocopper |
SMILES | [O--].[Cu++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11438510
|
Thermo Scientific Alfa Aesar
044356.06 |
5 g |
1088.71 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448510
|
Thermo Scientific Alfa Aesar
044356.14 |
25 g |
3810.47 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428510
|
Thermo Scientific Alfa Aesar
044356.22 |
100 g |
11265.74 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Copper(II) oxide is used as an adsorbent and absorbent, pigment, antifouling agent, wood preservative, optical glass polishing agent, welding flux for bronze materials, agricultural chemical and abrasive. It is used in the preparation of intermediates, oxidizing/reducing agents, surface treating agents and process regulators. It finds use in the manufacture of rayons, phosphors, and dry battery cells. It serves as a catalyst in certain chemical reactions of industrial importance. It is also employed in chemical sensors and superconducting materials. Hazardous materials like cyanides, dioxins and halogenated hydrocarbons can be disposed through oxidation with copper oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1317-38-0 | |
79.55 | |
KKCXRELNMOYFLS-UHFFFAOYSA-N | |
14829 | |
[O--].[Cu++] |
CuO | |
MFCD00010979 | |
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide | |
oxocopper |
Specifications
1326°C | |
Black | |
99.9995% (Metal basis) | |
MFCD00010979 | |
Practically insoluble in water,and alcohol. Soluble in dilute acids,alkali cyanides,and ammonium carbonate solution. | |
[O--].[Cu++] | |
79.55 | |
79.55 | |
2.63 | |
6.315 g/cm3 |
1317-38-0 | |
Powder | |
CuO | |
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide | |
KKCXRELNMOYFLS-UHFFFAOYSA-N | |
oxocopper | |
14829 | |
Odorless | |
Black powder | |
Copper(II) oxide |
Safety and Handling
H400-H410-H302: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.
missing translation for 'dotInformation' : DOT Class: 9, Packing Group: III
missing translation for 'einecsNumber' : 215-269-1
missing translation for 'rtecsNumber' : GL7900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only