Learn More
Masterflex™ Extension Cable, RJ-12
Make the most of your Masterflex™ pump with connection cables, replacement parts, and other accessories.
Brand: Masterflex™ 77095-04
1914.25 SEK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- Choose accessories and replacement parts suitable for your Masterflex™ pump
- Increase flexibility with extended cable lengths
Specifications
Optional extension cable, RJ12; 15 ft (4.5 m). | |
15 ft. | |
4.5 m |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
1Hours
4Minutes
37Seconds
37