Learn More
Corning™ Lambda Plus Pipettor Starter Kit
Designed for laboratories which are about to replace their old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes
Brand: Corning™ 4069
8315.96 SEK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
The Lambda Plus Starter Kit is designed for laboratories which are about to replace their old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes.
The Lambda Plus Starter Kit contains: four Lambda Plus Single-Channel pipettors conveniently packaged with useful accessories (manual, calibration key, color-coded rings, tube of piston grease and identification stickers).
- Lambda Plus Single-Channel pipettor:
- 0.5 to 10 μL (Cat. No. 4071)
- 2 to 20 μL (Cat. No. 4072)
- 20 to 200 μL (Cat. No. 4074)
- 100 to 1000 μL (Cat. No. 4075) - Universal carousel rack for six single-channel pipettors
- Corning DeckWorks™ Pipet Tips:
- 10 μL (Cat. No. 4135)
- 200 μL (Cat. No. 4138)
- 1000 μL (Cat. No. 4140) - Free Gift - 1 Corning insulated lunch bag
Specifications
1 | |
Lambda Plus | |
Starter Kit | |
Four single channel pipettors, manual, calibration key, color coded rings, tube of piston grease, identification stickers |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%