Learn More
Corning™ Clear Polystyrene 96-Well Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
2150.86 SEK - 4010.00 SEK
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Binding Type | Not Treated |
Color | Clear |
Material | Polystyrene |
Product Code | Brand | Well Shape | Surface Treatment | Sterility | Lid | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Well Shape | Surface Treatment | Sterility | Lid | Price | Quantity & Availability | ||||||
10265362
![]() |
Corning™
3788 |
Round | Non-treated | Sterile | With Lid |
4010.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||||
10367841
![]() |
Corning™
3896 |
Conical Bottom (V-well) | Non-treated | Sterile | Without Lid | N/A | N/A | N/A | |||||
10035351
![]() |
Corning™
3798 |
Round | Non-treated | Non-sterile | Without Lid |
3203.49 SEK
Pack of 100 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
10296381
![]() |
Corning™
3367 |
Round | Non-treated | Sterile | Without Lid |
2150.86 SEK
Pack of 50 |
In Stock
Log in to see stock. |
||||||
10163711
![]() |
Corning™
3795 |
Round | Non-treated | Sterile | Without Lid |
2983.31 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||||
10766482
![]() |
Corning™
3897 |
Conical Bottom (V-well) | Non-treated | Non-sterile | Without Lid |
3806.40 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||||
10216341
![]() |
Corning™
3370 |
Flat Bottom (F-well) | Non-treated | Sterile | With Lid |
3718.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||||
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Standard | |
Not Treated | |
Polystyrene | |
75 to 200 μL |
8 x 12 | |
Clear | |
96 |