Learn More
Corning™ Costar™ Thermowell™ Plate Sealing Mat
For use with Thermowell P, M, and all Thermowell 96-well polypropylene plates.
Brand: Corning™ 6555
This item is not returnable.
View return policy
Description
- For 96-well plates
- Fits Thermowell P (Cat. No. 07-200-541), Thermowell M (Cat. Nos. 07-200-245 and -691) and all Thermowell 96-well polypropylene plates
- Minimizes evaporation, condensation, and prevents cross-contamination
- Eliminates need for oil overlays in thermal cyclers with heated lids
- Reusable silicone rubber
- Autoclavable
Specifications
Silicone Rubber | |
Autoclavable | |
25 | |
Sealing Mat |
Non-sterile | |
96 | |
1 | |
96-well Polypropylene Plate |