Learn More
Fisherbrand™ Crimping Heads for Pneumatic Hand-held Crimping Tool
For use with Pneumatic Hand-held Crimping Tool.
5410.08 SEK - 10850.00 SEK
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
11865050
|
Fisherbrand™
|
20mm Crimping Head | Aluminum Cap |
5875.00 SEK
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11879440
|
Fisherbrand™
|
32mm Crimping Head | Flip Top/Flip Off Seals |
10850.00 SEK
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
12980981
|
Fisherbrand™
|
20mm Crimping Head | Flip Top/Flip Off Seals |
5865.00 SEK
Each |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11824970
|
Fisherbrand™
|
13mm Crimping Head | Flip Top/Flip Off Seals |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11869440
|
Fisherbrand™
|
32mm Crimping Head | Aluminum Cap |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11898951
|
Fisherbrand™
|
7.5mm Crimping Head | Aluminum/Steel Caps |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11868981
|
Fisherbrand™
|
8mm Crimping Head | Aluminum Cap |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11844890
|
Fisherbrand™
|
11mm Crimping Head | Aluminum Cap |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Crimping Head
- Pneumatic
- Steel