missing translation for 'onlineSavingsMsg'
Learn More

Crotonic Anhydride, MP Biomedicals

Product Code. 11432885
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 623-68-7
Molecular Formula C8H10O3
Quantity 10 g
Synonym crotonic anhydride, 2-butenoic anhydride, 2-butenoic acid, anhydride, crotonic acid anhydride, but-2-enoic anhydride, unii-n521pmq949, anhydrid kyseliny krotonove czech, crotonicanhydride, e-but-2-enoyl e-but-2-enoate, anhydrid kyseliny krotonove
InChI Key VJDDQSBNUHLBTD-GGWOSOGESA-N
SMILES CC=CC(=O)OC(=O)C=CC
IUPAC Name [(E)-but-2-enoyl] (E)-but-2-enoate
Molecular Weight (g/mol) 154.165
PubChem CID 6049834
Formula Weight 154.17
Chemical Name or Material Crotonic Anhydride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'