Learn More
BD Biosciences™ CS&T IVD Beads (CE/IVD)
Used to set up your cytometer, perform daily quality control, and determine lyse/wash application settings.
1955.00 SEK - 4835.00 SEK
Specifications
Product Type | Fluorescent Polystyrene Beads |
---|---|
Stability | The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 8 hours at 2°C–25°CC |
Product Code | Brand | Content And Storage | No. of Tests | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Content And Storage | No. of Tests | Price | Quantity & Availability | ||||
15846339
|
BD Biosciences™
656046 |
Store vials at 2°C–8°C and protect from light. | 50 Tests |
1955.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15856339
|
BD Biosciences™
656047 |
Three 3 mL vials at 50 tests per vial for 150 tests per kit | 150 Tests |
4835.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Intended for in vitro diagnostic use on BD FACSCanto™ II flow cytometers running BD FACSDiva™ software.
- Used to set up the cytometer, to perform daily performance quality control (QC), and to determine lyse/wash (LW) application settings.
- Dyed with fluorochromes that are excited by the cytometer's lasers.
Specifications
Fluorescent Polystyrene Beads | |
The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 8 hours at 2°C–25°CC |