Learn More
Sartorius Cubis™ II Micro Lab Balance
Offers readability of 1 ÎĽg and a maximum capacity between 3.1 g and 10.1 g. Used to determine the exact weight of very small sample quantities.
Brand:  Sartorius MCA3.6P-2S00-M QP99
 View more versions of this product
Product Code. 16393287
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The New Generation of Modular Premium Laboratory Balances
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series from Sartorius.
All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Balance with Type Approval Certificate for China
Color Touchscreen
Internal Calibration
5 sec.
80 mm
1.1 g, 2.1 g, 3.1 g
0.001 mg, 0.002 mg, 0.005 mg
Balance with Draft Shield M
Microbalance
100/240 V (± 10%), 50/60 Hz, 1.0 A
Videos
  • Sartorius - Cubis analytical balances
    Offers the optimal basis for users who need to weigh with the highest accuracy, but have lower needs in terms of complexity of a weighing process and in data management.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Sartorius Cubis analytical balances
    Product Title
    Sartorius Cubis™ II Micro Lab Balance > Capacity: 1.1 g, 2.1 g, 3.1 g; Readability: 0.001 mg, 0.002 mg, 0.005 mg; Balance with Draft Shield M