Learn More
Thermo Scientific™ CultiMaxx™ Shelving System for G-Rex 500M-CS Bioreactors
CultiMaxx™ Shelving Systems to support G-Rex 500M-CS Bioreactors
85678.03 SEK
Product Code | Brand | Description | For Use With (Equipment) | Includes | No. of Shelves | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Includes | No. of Shelves | Price | Quantity & Availability | ||
17988084
|
Thermo Scientific™
332395G08 |
Shelving system to support G-Rex® 500M for Reach-In 821L CO2 Incubator | Reach-In 821L CO2 Incubator | 7 levels of shelving | 7 |
85678.03 SEK
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Maximize the capacity, increase throughput and optimize cell growth in your lab using the CultiMaxx shelving system in a range of large capacity CO2 Incubators.
Maximize incubator space
- These shelving systems are designed to support scale-out of the G-Rex 500M-CS* in cell therapy production.
- With an in-depth review of the shelving design, this new system provides up to a 150% increase in capacity for the G-Rex, or another way to look at it: 2 1/2 incubators in 1.
Easy installation
- Shelving systems are retrofittable and can be self-installed.