Learn More
Fisherbrand™ Culture Tubes
Ideal for tissue culture, bacteriology, clinical chemistry, blood typing and cross matching procedures
Brand: Fisherbrand™ 73750-13100
4479.25 SEK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
Available with or without marking spot. Tubes packed in shrink wrapped trays. Ideal for tissue culture, bacteriology, clinical chemistry, blood typing and cross matching procedures.
Specifications
Test Tube | |
Borosilicate Glass | |
Tissue culture, bacteriology, clinical chemistry, blood typing and cross matching procedures. | |
1000 Pack | |
Not Included |
100 mm | |
Single-use | |
Tray; Shrink wrapped trays | |
13 mm |
Special Offers
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now