Learn More
Ambion™ Customer-Defined Pre-miR™ miRNA Precursor
Customer-Defined Pre-miR™ miRNA Precursor
Brand: Ambion™ AM17103
Additional Details : Weight : 0.01000kg
Description
Pre-miR™ miRNA Precursor Molecules are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs. Use our customization tool to design the ideal mimic for your miRNA sequence, or select from among predesigned products.Specifications
Room temperature | |
Pre-miRNA Precursor, 20nmol, Small, Chemically Modified Double-Stranded RNA Molecules Designed to Mimic Endogenous Mature miRNAs, Use Our Customization Tool to Design the Ideal Mimic for Your miRNA Sequence | |
Pre-miRNA Precursor | |
20 nmol |
For Research Use Only. Not for use in diagnostic procedures.