missing translation for 'onlineSavingsMsg'
Learn More

CVT-12012, MedChemExpress

MedChemExpress CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM, 6.1 nM for rat microsomal and human HEPG2, respectively.

Encompass_Preferred

1602.75 SEK - 9647.57 SEK

Specifications

Chemical Name or Material CVT-12012
Physical Form Powder
CAS 1018675-35-8
Color Yellow
Purity Grade Notes Research
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16475276
Encompass_Preferred
View Documents
Medchem Express
HY-11034
100 mg
9647.57 SEK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16467097
Encompass_Preferred
View Documents
Medchem Express
HY-11034
5 mg
1602.75 SEK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16453056
Encompass_Preferred
View Documents
Medchem Express
HY-11034
25 mg
4341.41 SEK
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16450697
Encompass_Preferred
View Documents
Medchem Express
HY-11034
50 mg
6504.33 SEK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16414378
Encompass_Preferred
View Documents
Medchem Express
HY-11034
10 mg
2411.90 SEK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM, 6.1 nM for rat microsomal and human HEPG2, respectively.
Specifications

Specifications

CVT-12012
1018675-35-8
Research
Metabolism-sugar/lipid metabolism
O=C(NCCN1C(C(C)=NC2=C1C=C(NCC3=CC=CC(C(F)(F)F)=C3)C=C2)=O)CO
434.41
Research
Powder
Yellow
C21H21F3N4O3
DMSO : 125 mg/mL (287.75 mM; Need ultrasonic)
434.41
99.16%
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
CVT-12012, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.