Learn More
CXCL10 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
2205.00 SEK - 19580.00 SEK
Specifications
Antigen | CXCL10 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17823613
|
Invitrogen
500-P93-50UG |
50 μg |
2205.00 SEK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17893213
|
Invitrogen
500-P93-100UG |
100 μg |
2830.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17863163
|
Invitrogen
500-P93-1MG |
1 mg |
19580.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: VPLSRTVRCT CISISNQPVN PRSLEKLEII PASQFCPRVE IIATMKKKGE KRCLNPESKA IKNLLKAVSK ERSKRSP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IP-10 (CXCL10). Anti-Human IP-10 (CXCL10)-specific antibody was purified by affinity chromatography employing an immobilized Human IP-10 (CXCL10) matrix. Sandwich ELISA: To detect Human IP-10 (CXCL10) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IP-10 (CXCL10) (500-P93Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IP-10 (CXCL10). Western Blot: To detect hIP-10 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIP-10 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.Specifications
CXCL10 | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P02778 | |
3627 | |
E.coli-derived Recombinant Human IP-10 (CXCL10) | |
Antigen affinity chromatography | |
CXCL10 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
10 kDa interferon gamma-induced protein; 10 kDa interferon-gamma induced protein precursor; C Cmotif chemokine; C X C motif chemokine; C7; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 10; chemokine C-X-C motif ligand 10; chemokine CXC-like protein; Crg2; CRG-2; CXC; CXC chemokine; CXC motif chemokine; C-X-C motif chemokine 10; C-X-C motif chemokine 10-like protein; C-X-C motif chemokine ligand 10; CXCL; CXCL10; CXCL10(1-73); gamma interferon inducible protein 10; gamma IP10; gamma-IP10; gIP-10; H-IP-10; IFI10; Inp10; interferon activated gene 10; interferon-gamma induced protein CRG-2; interferon-gamma-induced protein CRG-2; interferon-gamma-inducible protein 10; interferon-gamma-inducible protein-10; interferon-inducible cytokine IP-10; interferon-inducible protein 10; IP10; IP-10; M-IP-10; Mob1; mob-1; protein 10 from interferon (gamma)-induced cell line; protein Mob-1; Scyb10; similar to Small inducible cytokine B10 precursor (CXCL10) (10 kDa interferon-gamma induced protein) (Gamma-IP10) (IP-10); small inducible cytokine B subfamily (Cys-X-Cys) member 10; small inducible cytokine B subfamily (Cys-X-Cys), member 10; small inducible cytokine subfamily B (Cys-X-Cys), member 10; small-inducible cytokine B10 | |
CXCL10 | |
Primary | |
-20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.