missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL13 Monoclonal Antibody (143614), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA523871
This item is not returnable.
View return policy
Description
In ELISAs, this antibody does not cross-react with recombinant human (rh) CXCL13, rmCXCL11/I-TAC, rmCXCL9/MIG, or rm CXCL12/SDF-1 alpha. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CXCL13 Monoclonal antibody specifically detects CXCL13 in Mouse samples. It is validated for ELISA, Neutralization.Specifications
CXCL13 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O55038 | |
Cxcl13 | |
E. coli-derived recombinant mouse CXCL13 Ile22-Ala109 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
ELISA, Neutralization | |
143614 | |
Unconjugated | |
Cxcl13 | |
4631412M08Rik; Angie; ANGIE2; B cell-attracting chemokine 1; b lymphocyte chemoattractant; BCA1; BCA-1; B-cell chemoattractant; B-cell-attracting chemokine 1; B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1); BLC; BLR1L; B-lymphocyte chemoattractant; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chem; chemokine (C-X-C motif) ligand 13; chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant); CXC; CXC chemokine BLC; CXC motif chemokine; CXC motif chemokine 13; C-X-C motif chemokine 13; C-X-C motif chemokine ligand 13; CXCL; Cxcl13; SCYB13; small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cell chemoattractant); small inducible cytokine subfamily B (Cys-X-Cys), member 13; small-inducible cytokine B13 | |
Rat | |
Protein A/G | |
RUO | |
55985 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |